(1-Phenyl-1H-pyrazol-3-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A1186686-100MG A1186686-250MG A1186686-1G
76676-724EA 115.64 USD
76676-724 76676-726 77620-664
(1-Phenyl-1H-pyrazol-3-yl)methanamine hydrochloride 95%
(1-Phenyl-1H-pyrazol-3-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClN₃
MW: 209.6754 g/mol
MDL Number: MFCD12197097
CAS Number: 1170903-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR