3-Bromo-2-chloro-7-nitroquinoline 95%

Supplier: Ambeed
Warning

A1181203-100MG A1181203-1G A1181203-250MG
76676-702EA 48.81 USD
76676-702 76676-704 76676-706
3-Bromo-2-chloro-7-nitroquinoline 95%
3-Bromo-2-chloro-7-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₄BrClN₂O₂
MW: 287.4973 g/mol
MDL Number: MFCD18906541
CAS Number: 1026437-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR