2-(5-Bromo-1H-pyrrolo[2,3-b]pyridin-3-yl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
A1181195-100MG
A1181195-1G
76676-698EA
87
USD
76676-698
76676-700
2-(5-Bromo-1H-pyrrolo[2,3-b]pyridin-3-yl)acetic acid 98%
2-(5-Bromo-1H-pyrrolo[2,3-b]pyridin-3-yl)acetic acid
Precaución: for Research Use Only
Formula:
C₉H₇BrN₂O₂ MW: 255.07 g/mol |
MDL Number:
MFCD11100764 CAS Number: 1060795-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...