9-Mesityl-10-phenylacridin-10-ium chloride 96%

Supplier: Ambeed
Warning

A1180613-100MG A1180613-50MG
76676-694EA 84.88 USD
76676-694 76676-696
9-Mesityl-10-phenylacridin-10-ium chloride 96%
9-Mesityl-10-phenylacridin-10-ium chloride

Precaución: for Research Use Only

Formula: C₂₈H₂₄ClN
MW: 409.95 g/mol
MDL Number: MFCD32856356
CAS Number: 2088856-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR