(2-(4-Vinylphenyl)ethene-1,1,2-triyl)tribenzene 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A1180520-100MG A1180520-1G A1180520-250MG A1180520-5G
76676-682EA 14 USD
76676-682 76676-684 76676-686 76676-688
(2-(4-Vinylphenyl)ethene-1,1,2-triyl)tribenzene 98% stabilized
(2-(4-Vinylphenyl)ethene-1,1,2-triyl)tribenzene

Precaución: for Research Use Only

Formula: C₂₈H₂₂
MDL Number: MFCD32667035
CAS Number: 1351272-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR