6-Bromohexyl acrylate 98+% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A1180290-5G A1180290-25G A1180290-1G
76676-676EA 83 USD
76676-676 77571-076 77866-388
6-Bromohexyl acrylate 98+% stabilized
6-Bromohexyl acrylate

Precaución: for Research Use Only

Formula: C₉H₁₅BrO₂
MDL Number: MFCD32206237
CAS Number: 112231-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR