4,5-Dimethylphthalic acid 98%

Supplier: Ambeed
Warning

A1180062-1G A1180062-250MG A1180062-100MG
76676-660EA 92 USD
76676-660 76676-662 77866-385
4,5-Dimethylphthalic acid 98%
4,5-Dimethylphthalic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD20617334
CAS Number: 5680-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR