N-([1,1'-Biphenyl]-4-yl)-9-phenyl-9H-carbazol-3-amine 98%

Supplier: Ambeed
Warning

A1179875-1G A1179875-5G
76676-656EA 19.1 USD
76676-656 76676-658
N-([1,1'-Biphenyl]-4-yl)-9-phenyl-9H-carbazol-3-amine 98%
N-([1,1'-Biphenyl]-4-yl)-9-phenyl-9H-carbazol-3-amine

Precaución: for Research Use Only

Formula: C₃₀H₂₂N₂
MW: 410.5091 g/mol
CAS Number: 1002762-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR