9-(3-Bromophenyl)-9H-3,9'-bicarbazole 98%

Supplier: Ambeed
Warning

A1179873-1G A1179873-250MG A1179873-5G
76676-650EA 46.68 USD
76676-650 76676-652 76676-654
9-(3-Bromophenyl)-9H-3,9'-bicarbazole 98%
9-(3-Bromophenyl)-9H-3,9'-bicarbazole

Precaución: for Research Use Only

Formula: C₃₀H₁₉BrN₂
MW: 487.3893 g/mol
CAS Number: 1480890-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR