N1-(4-Bromophenyl)benzene-1,2-diamine 98%

Supplier: Ambeed
Warning

A1179870-1G A1179870-250MG
76676-642EA 36.08 USD
76676-642 76676-644
N1-(4-Bromophenyl)benzene-1,2-diamine 98%
N1-(4-Bromophenyl)benzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₂H₁₁BrN₂
MW: 263.13 g/mol
MDL Number: MFCD11201944
CAS Number: 100953-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR