1-(3,5,5,6,8,8-Hexamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one 98%

Supplier: Ambeed
Warning

A1179869-100G A1179869-25G A1179869-500G
76676-636EA 23.34 USD
76676-636 76676-638 76676-640
1-(3,5,5,6,8,8-Hexamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one 98%
1-(3,5,5,6,8,8-Hexamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₈H₂₆O
MW: 258.3985 g/mol
CAS Number: 1506-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR