5-Bromo-2-iodo-1,3-dimethylbenzene 98%

Supplier: Ambeed
Warning

A1179868-1G A1179868-5G
76676-632EA 18.04 USD
76676-632 76676-634
5-Bromo-2-iodo-1,3-dimethylbenzene 98%
5-Bromo-2-iodo-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrI
MW: 310.9576 g/mol
CAS Number: 260355-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR