Methyl 4-oxohexanoate 98%

Supplier: Ambeed
Warning

A1179863-1G A1179863-250MG
76676-628EA 105.03 USD
76676-628 76676-630
Methyl 4-oxohexanoate 98%
Methyl 4-oxohexanoate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.1684 g/mol
MDL Number: MFCD00069010
CAS Number: 2955-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR