(5AS,10bR)-2-(4-methoxyphenyl)-5a,10b-dihydro-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate ≥97%, ee 99%

Supplier: Ambeed
Danger

A1178609-100MG A1178609-250MG
76676-608EA 70.02 USD
76676-608 76676-610
(5AS,10bR)-2-(4-methoxyphenyl)-5a,10b-dihydro-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate ≥97%, ee 99%
(5AS,10bR)-2-(4-methoxyphenyl)-5a,10b-dihydro-4H,6H-indeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₉H₁₈BF₄N₃O₂
MW: 407.1697 g/mol
CAS Number: 463326-78-5
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR