4-(2-Fluoropyridin-4-yl)morpholine 97%

Supplier: Ambeed
Warning

A1178536-1G A1178536-250MG
76676-598EA 700.2 USD
76676-598 76676-600
4-(2-Fluoropyridin-4-yl)morpholine 97%
4-(2-Fluoropyridin-4-yl)morpholine

Precaución: for Research Use Only

Formula: C₉H₁₁FN₂O
MW: 182.1949 g/mol
MDL Number: MFCD26092229
CAS Number: 1564591-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR