9-((2R,4S,5R)-4-((tert-Butyldimethylsilyl)oxy)-5-(((tert-butyldimethylsilyl)oxy)methyl)tetrahydrofuran-2-yl)-9H-purin-6-amine 98%

Supplier: Ambeed
Warning

A1178101-100MG A1178101-1G A1178101-250MG
76676-592EA 36.08 USD
76676-592 76676-594 76676-596
9-((2R,4S,5R)-4-((tert-Butyldimethylsilyl)oxy)-5-(((tert-butyldimethylsilyl)oxy)methyl)tetrahydrofuran-2-yl)-9H-purin-6-amine 98%
9-((2R,4S,5R)-4-((tert-Butyldimethylsilyl)oxy)-5-(((tert-butyldimethylsilyl)oxy)methyl)tetrahydrofuran-2-yl)-9H-purin-6-amine

Precaución: for Research Use Only

Formula: C₂₂H₄₁N₅O₃Si₂
MW: 479.7637 g/mol
MDL Number: MFCD22373345
CAS Number: 51549-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR