Sodium hexachloropalladate(IV) 98%

Supplier: Ambeed
Warning

Synonyms: Disodium hexachloropalladate

A1177954-100MG A1177954-1G A1177954-250MG
76676-584EA 111.4 USD
76676-584 76676-586 76676-588
Sodium hexachloropalladate(IV) 98%
Sodium hexachloropalladate(IV)

Precaución: for Research Use Only

Formula: Cl₆Na₂Pd
MW: 365.12 g/mol
MDL Number: MFCD00014235
CAS Number: 53823-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR