Palladium(II) benzoate 99%

Supplier: Ambeed
Warning

A1177935-100MG A1177935-1G A1177935-250MG
76676-578EA 145 USD
76676-578 76676-580 76676-582
Palladium(II) benzoate 99%
Palladium(II) benzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄Pd
MDL Number: MFCD18802284
CAS Number: 3375-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR