3,4,5-Trimethylthiazol-3-ium iodide 97%

Supplier: Ambeed
Warning

A1177914-1G A1177914-250MG A1177914-5G A1177914-100MG
76676-572EA 45 USD
76676-572 76676-574 76676-576 77866-366
3,4,5-Trimethylthiazol-3-ium iodide 97%
3,4,5-Trimethylthiazol-3-ium iodide

Precaución: for Research Use Only

Formula: C₆H₁₀INS
MDL Number: MFCD00498005
CAS Number: 62993-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR