(11bS)-N-Benzhydryl-N-isopropyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine 95%

Supplier: Ambeed
Warning

A1177903-100MG A1177903-1G A1177903-250MG
76676-562EA 49.87 USD
76676-562 76676-564 76676-566
(11bS)-N-Benzhydryl-N-isopropyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine 95%
(11bS)-N-Benzhydryl-N-isopropyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine

Precaución: for Research Use Only

Formula: C₃₆H₃₀NO₂P
MW: 539.6027 g/mol
MDL Number: MFCD31707590
CAS Number: 1637749-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR