(11bR)-4-Hydroxy-2,6-bis(4-methoxyphenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%

Supplier: Ambeed
Warning

A1177896-100MG A1177896-1G A1177896-250MG
76676-552EA 41 USD
76676-552 76676-554 76676-556
(11bR)-4-Hydroxy-2,6-bis(4-methoxyphenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%
(11bR)-4-Hydroxy-2,6-bis(4-methoxyphenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₃₄H₃₃O₆P
CAS Number: 1011465-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR