1,3-Dimesityl-1H-imidazol-3-ium hydrogen carbonate 98%

Supplier: Ambeed
Danger

A1177874-1G A1177874-250MG A1177874-5G
76676-534EA 79.57 USD
76676-534 76676-536 76676-538
1,3-Dimesityl-1H-imidazol-3-ium hydrogen carbonate 98%
1,3-Dimesityl-1H-imidazol-3-ium hydrogen carbonate

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₂O₃
MW: 366.4535 g/mol
MDL Number: MFCD28400093
CAS Number: 1372124-93-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR