(3AR,3a'R,8aS,8a'S)-2,2'-(cycloheptane-1,1-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) ≥98%, ee 99%

Supplier: Ambeed
Warning

A1177860-100MG A1177860-1G A1177860-250MG
76676-524EA 111.4 USD
76676-524 76676-526 76676-528
(3AR,3a'R,8aS,8a'S)-2,2'-(cycloheptane-1,1-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) ≥98%, ee 99%
(3AR,3a'R,8aS,8a'S)-2,2'-(cycloheptane-1,1-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole)

Precaución: for Research Use Only

Formula: C₂₇H₂₈N₂O₂
MW: 412.5234 g/mol
MDL Number: MFCD32201213
CAS Number: 2085239-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR