(4S,4'S)-2,2'-(Cyclohexane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) ≥97%, ee 99%

Supplier: Ambeed
Warning

A1177859-250MG A1177859-100MG A1177859-1G
76676-522EA 130 USD
76676-522 77620-598 77866-363
(4S,4'S)-2,2'-(Cyclohexane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) ≥97%, ee 99%
(4S,4'S)-2,2'-(Cyclohexane-1,1-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₀H₃₄N₂O₂
MW: 334.5 g/mol
MDL Number: MFCD32201212
CAS Number: 298693-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR