Ethyl 2-(2-(4,5-bis(4-methoxyphenyl)thiazol-2-yl)-1H-pyrrol-1-yl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Pamicogrel

A1177666-1MG A1177666-5MG
76676-488EA 13.8 USD
76676-488 76676-490
Ethyl 2-(2-(4,5-bis(4-methoxyphenyl)thiazol-2-yl)-1H-pyrrol-1-yl)acetate 98%
Ethyl 2-(2-(4,5-bis(4-methoxyphenyl)thiazol-2-yl)-1H-pyrrol-1-yl)acetate

Precaución: for Research Use Only

Formula: C₂₅H₂₄N₂O₄S
MW: 448.5341 g/mol
MDL Number: MFCD00866172
CAS Number: 101001-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR