N2-Cyclopropyl-N4-(1-isopropylpiperidin-4-yl)-6,7-dimethoxyquinazoline-2,4-diamine 98%

Supplier: Ambeed
Warning

A1177125-100MG A1177125-10MG A1177125-1MG A1177125-50MG A1177125-5MG A1177125-250MG
76676-470EA 238 USD
76676-470 76676-472 76676-474 76676-476 76676-478 77866-343
N2-Cyclopropyl-N4-(1-isopropylpiperidin-4-yl)-6,7-dimethoxyquinazoline-2,4-diamine 98%
N2-Cyclopropyl-N4-(1-isopropylpiperidin-4-yl)-6,7-dimethoxyquinazoline-2,4-diamine

Precaución: for Research Use Only

Formula: C₂₁H₃₁N₅O₂
MDL Number: MFCD32197197
CAS Number: 2328073-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR