DMNQ 99%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dimethoxy-1,4-naphthoquinone, 2,3-Dimethoxynaphthalene-1,4-dione

A1177106-10MG A1177106-1MG A1177106-5MG
76676-464EA 181.42 USD
76676-464 76676-466 76676-468
DMNQ 99%
DMNQ

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₄
MW: 218.2 g/mol
MDL Number: MFCD00184331
CAS Number: 6956-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR