4-(Pyridin-3-yl)-4,5-dihydropyrrolo[1,2-a]quinoxaline 98%

Supplier: Ambeed
Warning

Synonyms: UBCS039

A1177029-10MG A1177029-1MG A1177029-50MG A1177029-5MG
76676-440EA 29.71 USD
76676-440 76676-442 76676-444 76676-446
4-(Pyridin-3-yl)-4,5-dihydropyrrolo[1,2-a]quinoxaline 98%
4-(Pyridin-3-yl)-4,5-dihydropyrrolo[1,2-a]quinoxaline

Precaución: for Research Use Only

Formula: C₁₆H₁₃N₃
MW: 247.2945 g/mol
MDL Number: MFCD00206184
CAS Number: 358721-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR