rel-4-(((1R,4R)-4-hydroxycyclohexyl)amino)-4'-isopropyl-[1,1'-biphenyl]-3-carboxamide 98%

Supplier: Ambeed
Warning

A1176950-10MG A1176950-1MG A1176950-50MG A1176950-5MG
76676-426EA 78.51 USD
76676-426 76676-428 76676-430 76676-432
rel-4-(((1R,4R)-4-hydroxycyclohexyl)amino)-4'-isopropyl-[1,1'-biphenyl]-3-carboxamide 98%
rel-4-(((1R,4R)-4-hydroxycyclohexyl)amino)-4'-isopropyl-[1,1'-biphenyl]-3-carboxamide

Precaución: for Research Use Only

Formula: C₂₂H₂₈N₂O₂
MW: 352.47 g/mol
MDL Number: MFCD32067891
CAS Number: 2234897-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR