tert-Butyl 4-(4-(2-(3-methyl-4-oxo-3,4-dihydrophthalazin-1-yl)acetamido)benzyl)piperazine-1-carboxylate 99+%

Supplier: Ambeed
Warning

A1176916-10MG A1176916-1MG A1176916-25MG A1176916-50MG A1176916-5MG
76676-414EA 395.72 USD
76676-414 76676-416 76676-418 76676-420 76676-422
tert-Butyl 4-(4-(2-(3-methyl-4-oxo-3,4-dihydrophthalazin-1-yl)acetamido)benzyl)piperazine-1-carboxylate 99+%
tert-Butyl 4-(4-(2-(3-methyl-4-oxo-3,4-dihydrophthalazin-1-yl)acetamido)benzyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₇H₃₃N₅O₄
MW: 491.5820 g/mol
MDL Number: MFCD22666400
CAS Number: 1311174-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR