N4-((4-(Aminomethyl)cyclohexyl)methyl)-5-nitro-N2-(2-(trifluoromethoxy)benzyl)pyrimidine-2,4-diamine 99%

Supplier: Ambeed
Warning

A1176892-10MG A1176892-1MG A1176892-2MG A1176892-5MG
76676-406EA 42 USD
76676-406 76676-408 76676-410 76676-412
N4-((4-(Aminomethyl)cyclohexyl)methyl)-5-nitro-N2-(2-(trifluoromethoxy)benzyl)pyrimidine-2,4-diamine 99%
N4-((4-(Aminomethyl)cyclohexyl)methyl)-5-nitro-N2-(2-(trifluoromethoxy)benzyl)pyrimidine-2,4-diamine

Precaución: for Research Use Only

Formula: C₂₀H₂₅F₃N₆O₃
MDL Number: MFCD31715442
CAS Number: 736048-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR