(E)-N-(4-((E)-3,5-Dimethoxystyryl)phenyl)-3-(3,4,5-trimethoxyphenyl)acrylamide 98%

Supplier: Ambeed
Warning

A1176397-10MG A1176397-1MG A1176397-25MG A1176397-50MG A1176397-5MG
76676-374EA 22 USD
76676-374 76676-376 76676-378 76676-380 76676-382
(E)-N-(4-((E)-3,5-Dimethoxystyryl)phenyl)-3-(3,4,5-trimethoxyphenyl)acrylamide 98%
(E)-N-(4-((E)-3,5-Dimethoxystyryl)phenyl)-3-(3,4,5-trimethoxyphenyl)acrylamide

Precaución: for Research Use Only

Formula: C₂₈H₂₉NO₆
MDL Number: MFCD32174234
CAS Number: 2059952-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR