1-Butylpyridiniumtrifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Butylpyridin-1-ium trifluoromethanesulphonate

A1176322-1G A1176322-25G A1176322-5G
76676-368EA 7.43 USD
76676-368 76676-370 76676-372
1-Butylpyridiniumtrifluoromethanesulfonate 98%
1-Butylpyridiniumtrifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₁₀H₁₄F₃NO₃S
MW: 285.29 g/mol
MDL Number: MFCD03427622
CAS Number: 390423-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR