3-Benzyl-1,2-dimethyl-1H-imidazol-3-ium tetrafluoroborate 98%

Supplier: Ambeed
Warning

A1176220-25G A1176220-5G
76676-362EA 60.48 USD
76676-362 76676-364
3-Benzyl-1,2-dimethyl-1H-imidazol-3-ium tetrafluoroborate 98%
3-Benzyl-1,2-dimethyl-1H-imidazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₄N₂
MW: 274.0655 g/mol
MDL Number: MFCD30187962
CAS Number: 862999-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR