1-(4-sulfobutyl)pyridin-1-ium hydrogen sulfate 97%

Supplier: Ambeed
Danger

A1176212-25G A1176212-1G
76676-360EA 49 USD
76676-360 77866-276
1-(4-sulfobutyl)pyridin-1-ium hydrogen sulfate 97%
1-(4-sulfobutyl)pyridin-1-ium hydrogen sulfate

Precaución: for Research Use Only

Formula: C₉H₁₅NO₇S₂
MW: 313.3479 g/mol
MDL Number: MFCD30187696
CAS Number: 827320-61-6
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR