7-Methyl-2-((7-methyl-[1,2,4]triazolo[1,5-a]pyridin-6-yl)amino)-9-(tetrahydro-2H-pyran-4-yl)-7H-purin-8(9H)-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
AZD-7648
A1175994-10MG
A1175994-1MG
A1175994-5MG
A1175994-50MG
76676-354EA
114
USD
76676-354
76676-356
76676-358
77620-488
7-Methyl-2-((7-methyl-[1,2,4]triazolo[1,5-a]pyridin-6-yl)amino)-9-(tetrahydro-2H-pyran-4-yl)-7H-purin-8(9H)-one 98%
7-Methyl-2-((7-methyl-[1,2,4]triazolo[1,5-a]pyridin-6-yl)amino)-9-(tetrahydro-2H-pyran-4-yl)-7H-purin-8(9H)-one
Precaución: for Research Use Only
Formula:
C₁₈H₂₀N₈O₂ |
MDL Number:
MFCD32062688 CAS Number: 2230820-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...