Bis [2- (2,4-difluorophenyl) -5-trifluoromethylpyridine] [1,10-phenanthroline] iridium hexafluorophosphate 98%

Supplier: Ambeed
Warning

A1175915-100MG A1175915-1G A1175915-250MG A1175915-50MG
76676-340EA 71 USD
76676-340 76676-342 76676-344 76676-346
Bis [2- (2,4-difluorophenyl) -5-trifluoromethylpyridine] [1,10-phenanthroline] iridium hexafluorophosphate 98%
Bis [2- (2,4-difluorophenyl) -5-trifluoromethylpyridine] [1,10-phenanthroline] iridium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₆H₁₈F₁₆IrN₄P
CAS Number: 1639408-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR