[5,5'-Bis(trifluoromethyl)-2,2'-bipyridine-κN,κN]bis[3,5-difluoro-2-[5-(trifluoromethyl)-2-pyridinyl-κN]phenyl] iridium hexafluorophosphate 98%

Supplier: Ambeed
Warning

A1175854-100MG A1175854-1G A1175854-250MG A1175854-50MG A1175854-5G
76676-324EA 81.69 USD
76676-324 76676-326 76676-328 76676-330 76676-332
[5,5'-Bis(trifluoromethyl)-2,2'-bipyridine-κN,κN]bis[3,5-difluoro-2-[5-(trifluoromethyl)-2-pyridinyl-κN]phenyl] iridium hexafluorophosphate 98%
[5,5'-Bis(trifluoromethyl)-2,2'-bipyridine-κN,κN]bis[3,5-difluoro-2-[5-(trifluoromethyl)-2-pyridinyl-κN]phenyl] iridium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₆H₁₆F₂₂IrN₄P
MW: 1145.6947 g/mol
MDL Number: MFCD31707657
CAS Number: 1973375-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR