4,4'-Bis(trifluoromethyl)-2,2'-bipyridinebis[3,5-difluoro-2-[5-trifluoromethyl-2-pyridinyl)phenyl] iridium(III) hexafluorophosphate 97%

Supplier: Ambeed
Warning

A1175853-100MG A1175853-1G A1175853-250MG A1175853-50MG
76676-316EA 80 USD
76676-316 76676-318 76676-320 76676-322
4,4'-Bis(trifluoromethyl)-2,2'-bipyridinebis[3,5-difluoro-2-[5-trifluoromethyl-2-pyridinyl)phenyl] iridium(III) hexafluorophosphate 97%
4,4'-Bis(trifluoromethyl)-2,2'-bipyridinebis[3,5-difluoro-2-[5-trifluoromethyl-2-pyridinyl)phenyl] iridium(III) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₆H₁₆F₂₂IrN₄P
MDL Number: MFCD31707656
CAS Number: 2030437-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR