(S)-2-(2-(Diphenylphosphanyl)phenyl)-4-isobutyl-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1175344-100MG A1175344-50MG
76676-290EA 122.01 USD
76676-290 76676-292
(S)-2-(2-(Diphenylphosphanyl)phenyl)-4-isobutyl-4,5-dihydrooxazole 98%
(S)-2-(2-(Diphenylphosphanyl)phenyl)-4-isobutyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₅H₂₆NOP
MW: 387.4538 g/mol
CAS Number: 167693-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR