(S)-3-(tert-Butyl)-4-(4',6'-dimethoxy-[1,1':3',1''-terphenyl]-5'-yl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 97%

Supplier: Ambeed
Warning

A1175271-100MG A1175271-50MG
76676-288EA 164.44 USD
76676-288 77620-464
(S)-3-(tert-Butyl)-4-(4',6'-dimethoxy-[1,1':3',1''-terphenyl]-5'-yl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 97%
(S)-3-(tert-Butyl)-4-(4',6'-dimethoxy-[1,1':3',1''-terphenyl]-5'-yl)-2,3-dihydrobenzo[d][1,3]oxaphosphole

Precaución: for Research Use Only

Formula: C₃₁H₃₁O₃P
MW: 482.55 g/mol
MDL Number: MFCD32641163
CAS Number: 2021201-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR