(2S,3S)-3-(tert-butyl)-4-(2,6-dimethoxyphenyl)-2-ethyl-2,3-dihydrobenzo[d][1,3]oxaphosphole 97%

Supplier: Ambeed
Warning

A1175220-100MG A1175220-250MG
76676-266EA 113.52 USD
76676-266 77620-444
(2S,3S)-3-(tert-butyl)-4-(2,6-dimethoxyphenyl)-2-ethyl-2,3-dihydrobenzo[d][1,3]oxaphosphole 97%
(2S,3S)-3-(tert-butyl)-4-(2,6-dimethoxyphenyl)-2-ethyl-2,3-dihydrobenzo[d][1,3]oxaphosphole

Precaución: for Research Use Only

Formula: C₂₁H₂₇O₃P
MW: 358.4111 g/mol
CAS Number: 2247162-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR