(R)-3-(tert-butyl)-4-(2-methoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 95%

Supplier: Ambeed
Warning

A1175218-100MG A1175218-1G A1175218-250MG
76676-262EA 76 USD
76676-262 77866-253 77866-254
(R)-3-(tert-butyl)-4-(2-methoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 95%
(R)-3-(tert-butyl)-4-(2-methoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole

Precaución: for Research Use Only

Formula: C₁₈H₂₁O₂P
CAS Number: 1338454-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR