Cobalt(II) trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A1175154-1G A1175154-25G A1175154-5G A1175154-100G
76676-230EA 7 USD
76676-230 76676-232 76676-234 77596-028
Cobalt(II) trifluoromethanesulfonate 98%
Cobalt(II) trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₂CoF₆O₆S₂
MW: 357.0714 g/mol
MDL Number: MFCD31707617
CAS Number: 58164-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR