(2S,2'S,3S,3'S)-3,3'-Di-tert-butyl-2,2'-diisopropyl-2,2',3,3'-tetrahydro-4,4'-bibenzo[d][1,3]oxaphosphole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1175125-100MG A1175125-50MG A1175125-250MG
76676-222EA 309.79 USD
76676-222 76676-224 77620-432
(2S,2'S,3S,3'S)-3,3'-Di-tert-butyl-2,2'-diisopropyl-2,2',3,3'-tetrahydro-4,4'-bibenzo[d][1,3]oxaphosphole ≥97%, ee 99%
(2S,2'S,3S,3'S)-3,3'-Di-tert-butyl-2,2'-diisopropyl-2,2',3,3'-tetrahydro-4,4'-bibenzo[d][1,3]oxaphosphole

Precaución: for Research Use Only

Formula: C₂₈H₄₀O₂P₂
MW: 470.5635 g/mol
CAS Number: 2207601-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR