(3S,3'S)-3,3'-Di-tert-butyl-2,2',3,3'-tetrahydro-4,4'-bibenzo[d][1,3]oxaphosphole 97%

Supplier: Ambeed
Warning

A1175122-100MG A1175122-250MG A1175122-50MG
76676-216EA 276 USD
76676-216 77866-247 77866-248
(3S,3'S)-3,3'-Di-tert-butyl-2,2',3,3'-tetrahydro-4,4'-bibenzo[d][1,3]oxaphosphole 97%
(3S,3'S)-3,3'-Di-tert-butyl-2,2',3,3'-tetrahydro-4,4'-bibenzo[d][1,3]oxaphosphole

Precaución: for Research Use Only

Formula: C₂₂H₂₈O₂P₂
MDL Number: MFCD32201250
CAS Number: 2207601-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR