2-Hydroxy-2-phosphono-3-(pyridin-3-yl)propanoic acid 99%

Supplier: Ambeed
Warning

Synonyms: NE 10790

A1175086-10MG A1175086-1MG A1175086-50MG A1175086-5MG A1175086-1G A1175086-250MG A1175086-100MG
76676-206EA 106 USD
76676-206 76676-208 76676-210 76676-212 77596-024 77596-026 77620-428
2-Hydroxy-2-phosphono-3-(pyridin-3-yl)propanoic acid 99%
2-Hydroxy-2-phosphono-3-(pyridin-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₀NO₆P
MW: 247.14 g/mol
MDL Number: MFCD20233001
CAS Number: 152831-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR