3-Butyl-1-vinyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A1174281-5G A1174281-100G
76676-186EA 21 USD
76676-186 77866-245
3-Butyl-1-vinyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide 98% stabilized
3-Butyl-1-vinyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide

Precaución: for Research Use Only

Formula: C₁₁H₁₅F₆N₃O₄S₂
MDL Number: MFCD18251312
CAS Number: 758716-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR