N-(4-((4-Aminophenyl)thio)phenyl)-4-(4-methoxyphenylsulfonamido)butanamide 98%

Supplier: Ambeed
Warning

A1173077-10MG A1173077-1MG A1173077-5MG
76676-180EA 53 USD
76676-180 76676-182 76676-184
N-(4-((4-Aminophenyl)thio)phenyl)-4-(4-methoxyphenylsulfonamido)butanamide 98%
N-(4-((4-Aminophenyl)thio)phenyl)-4-(4-methoxyphenylsulfonamido)butanamide

Precaución: for Research Use Only

Formula: C₂₃H₂₅N₃O₄S₂
MW: 471.59 g/mol
MDL Number: MFCD07784498
CAS Number: 791835-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR