N1,N2-Bis(4-(pyridin-2-yl)benzyl)ethane-1,2-diamine 98%

Supplier: Ambeed
Warning

Synonyms: BC-1215

A1173075-10MG A1173075-1MG A1173075-25MG A1173075-50MG A1173075-5MG
76676-170EA 51.99 USD
76676-170 76676-172 76676-174 76676-176 76676-178
N1,N2-Bis(4-(pyridin-2-yl)benzyl)ethane-1,2-diamine 98%
N1,N2-Bis(4-(pyridin-2-yl)benzyl)ethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₂₆H₂₆N₄
MW: 394.51 g/mol
MDL Number: MFCD28118991
CAS Number: 1507370-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR